Halophilic Broth Dehydrated Culture Media, HiMedia Laboratories
95022-044
: HiMedia
:
- Description:Halophilic Broth
- Cat. No.:95022-044
- Supplier no.:M591-500G
For the isolation and cultivation of extremely halophilic bacteria
- Microbiological culture media
- 500 g poly bottles