High Plate Count Agar Dehydrated Culture Media, HiMedia
Catalog # 95020-472
Supplier: HiMedia
CAS Number:
Specifications
- Description:High Plate Count Agar
- Cat. No.:95020-472
- Supplier no.:M1097-500G
Specifications
About this item
Used for obtaining higher colony counts by spread plate or pour plate or membrane filter technique
- Microbiological culture media
- 500 g poly bottles