BD Difco™ Mycoplasma Supplement, BD Diagnostics
Catalog # 90004-046
Supplier: BD
CAS Number:
Specifications
- Description:Mycoplasma Supplement
- Size:6 x 30 mL
- Cat. No.:90004-046
- Supplier no.:283610
Specifications
About this item
These mycoplasma supplement are prepared from yeast extract and horse serum for the isolation and cultivation of Mycoplasma species
For use with PPLO agar and agar base, PPLO broth and broth base.