ContactUsLinkComponent
GluTracker™ Glucose Uptake Assay Kit (Cell-Based), BioVision Inc.
GluTracker™ Glucose Uptake Assay Kit (Cell-Based), BioVision Inc.
Catalog # 76022-218
Supplier:  Avantor
CAS Number:  
undefined
GluTracker™ Glucose Uptake Assay Kit (Cell-Based), BioVision Inc.
Catalog # 76022-218
Supplier:  Avantor
CAS Number:  
Specifications
  • Capacity:
    50 assays
  • Description:
    GluTracker™ Glucose Uptake Assay Kit
  • Size:
    50 assays
  • Brand Name:
    BioVision Inc.
  • Cat. No.:
    76022-218
Specifications
About this item
Non-radioactive method to image and accurately measure glucose uptake via FACS
  • Easy-to-use
  • Non-radioactive
  • Image and accurately measure glucose uptake in cultured cells in response to insulin, growth factors etc.
Glucose is a ubiquitous energy source in most organisms and plays a pivotal role in cellular metabolism and homeostasis. Cancer cells exhibit increased glucose uptake to support their high proliferation rate. At BioVision, we have developed a fluorescent glucose analog, which just like glucose can be taken up by cells through glucose transporters. However, this glucose analog cannot be fully utilized in glycolysis because of its modification and thus accumulates inside the cells. Fluorescence generated by this fluorescent glucose analog is proportional to the glucose uptake by the cells and can be used to measure glucose uptake using fluorescent microscopy and flow cytometry. To validate the assay, the kit includes phloretin, a natural phenol that inhibits glucose uptake. This easy-to-use, non-radioactive kit allows imaging and accurate measurement of glucose uptake in cultured cells in response to insulin, growth factors etc.