Malt Extract Broth, Modified as per Thom and Church, Dehydrated Culture Media, HiMedia
Catalog # 95020-532
Supplier: HiMedia
CAS Number:
Specifications
- Description:Malt Extract Broth, Modified as per Thom and Church
- Cat. No.:95020-532
- Supplier no.:M1128-500G
Specifications
About this item
For the cultivation and enumeration of yeasts and molds and to check sterility to detect presence of these organisms.
- Microbiological culture media
- 500 g poly bottles