MacConkey Agar with 0.15% Bile Salts, CV, and NaCl, Dehydrated Culture Media, HiMedia
Catalog # 95020-204
Supplier: HiMedia
CAS Number:
Specifications
- Description:MacConkey Agar with 0.15% Bile Salts, CV, and NaCl
- Cat. No.:95020-204
- Supplier no.:M081-500G
Specifications
About this item
For the selective isolation and differentiation of coliform organisms and other enteric pathogens.
- Microbiological culture media
- 500 g poly bottles