Fungal Broth with Low pH Dehydrated Culture Media, HiMedia
Catalog # 95021-576
Supplier: HiMedia
CAS Number:
Specifications
- Description:Fungal Broth with Low pH (Mycological Broth with Low pH)
- Cat. No.:95021-576
- Supplier no.:M265-500G
Specifications
About this item
For the selective enumeration and cultivation of saprophytic fungi and aciduric bacteria.
- Microbiological culture media
- 500 g poly bottles