Gelatin Phosphate Buffer Dehydrated Culture Media, HiMedia
Catalog # 95021-012
Supplier: HiMedia
CAS Number:
Specifications
- Description:Gelatin Phosphate Buffer
- Cat. No.:95021-012
- Supplier no.:M1359-500G
Specifications
About this item
For toxin detection in food products when Clostridium botulinum is suspected.
- Microbiological culture media
- 500 g poly bottles