Cell-Mate3D™ Cell Retrieval Kit, BRTI Life Sciences
Catalog # 76036-026
Supplier: BRTI LIFE SCIENCES
CAS Number:
Specifications
- Description:Cell-Mate3D™ Cell Retrieval Kit
- Size:3 Tubes - 15 Isolations
- Cat. No.:76036-026
Specifications
About this item
Isolate cells from Cell-Mate3D matrix simple 15 minutes process cancer, stem cells, normal cells.
- Up to 85% recovery of viable cells
- Simple and easy 15 minutes matrix digestion
- Use for flow cytrometry
- Sort specific cell populations