Culture Medium for RWC, Dehydrated, HiMedia Laboratories
95021-908
: HiMedia
:
- Description:Culture Medium for RWC (Disinfectant Test Broth, RWC Medium)
- Cat. No.:95021-908
- Supplier no.:M500-500G
For the determination of phenol coefficients of disinfectants using Salmonella typhi as a test organism.
- Microbiological culture media
- 500 g poly bottles