Columbia Blood Agar Base with 1% Agar, Dehydrated Culture Media, HiMedia
Catalog # 95021-142
Supplier: HiMedia
CAS Number:
Specifications
- Description:Columbia Blood Agar Base with 1% Agar
- Cat. No.:95021-142
- Supplier no.:M144A-500G
Specifications
About this item
Basal medium used with or without blood for the isolation and cultivation of fastidious bacteria.
- Microbiological culture media
- 500 g poly bottles