Dey-Engley Neutralizing Broth, without Bromocresol Purple, Dehydrated Culture Media, HiMedia Laboratories
Catalog # 95039-380
Supplier: HiMedia
CAS Number:
Specifications
- Description:Dey-Engley (D/E) Neutralizing Broth (without Bromocresol Purple)
- Cat. No.:95039-380
- Supplier no.:MU1062-500G
Specifications
About this item
For disinfectant testing, where neutralizaton of antiseptics and disinfectants is important for determining bactericidal activity, in accordance with USP.
- Microbiological culture media
- 500 g poly bottles