Enterococcus Confirmatory Agar Dehydrated Culture Media, HiMedia
95021-766
: HiMedia
:
- Description:Enterococcus Confirmatory Agar
- Cat. No.:95021-766
- Supplier no.:M392-500G
For confirming the presence of enterococci in water supplies and other sources.
- Microbiological culture media
- 500 g poly bottles