ContactUsLinkComponent
Access Key Options for Ultra-Low Temperature Freezers, Thermo Scientific
Access Key Options for Ultra-Low Temperature Freezers, Thermo Scientific
Catalog # 76019-738
undefined
Access Key Options for Ultra-Low Temperature Freezers, Thermo Scientific
Catalog # 76019-738
Supplier Number:  FAC34567LE
Specifications
  • Description:
    Access Key Options for TSX Series, Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE
  • Cat. No.:
    76019-738
Specifications
About this item
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Ordering information: Field-Installed access key option includes card access with five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.