Specifications
- Description:Tryptic Soy Agar with Polysorbate 80 and Lecithin
- Size:500 g
- Cat. No.:EM1.07324.0500
- Supplier no.:1.07324.0500
Specifications
About this item
Media is granulated.
- Granulation preserves homogeneity
- Easy to weigh out
- Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.