Order Entry
United States
ContactUsLinkComponent
Sabouraud 4% Dextrose Agar Dehydrated Culture Media, MilliporeSigma
Sabouraud 4% Dextrose Agar Dehydrated Culture Media, MilliporeSigma
Catalog # EM1.05438.0500
Supplier:  MilliporeSigma
CAS Number:  
undefined
Sabouraud 4% Dextrose Agar Dehydrated Culture Media, MilliporeSigma
Catalog # EM1.05438.0500
Supplier:  MilliporeSigma
Supplier Number:  1.05438.0500
CAS Number:  

Specifications

  • Description:
    Sabouraud 4%-Dextrose Agar
  • Size:
    500 g
  • Cat. No.:
    EM1.05438.0500
  • Supplier no.:
    1.05438.0500

Specifications

About this item

Media is granulated.

  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media

Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.