Order Entry
United States
ContactUsLinkComponent
Screen-Well® Myotoxicity Library, Enzo Life Sciences
Screen-Well® Myotoxicity Library, Enzo Life Sciences
Catalog # 76003-308
Supplier:  Enzo Life Sciences
CAS Number:  
Screen-Well® Myotoxicity Library, Enzo Life Sciences
Catalog # 76003-308
Supplier:  Enzo Life Sciences
Supplier Number:  LIB101-0100
CAS Number:  

Specifications

  • Description:
    1 Library
  • Size:
    100 µL/Well
  • Concentration:
    DMSO solutions (10mM)
  • Cat. No.:
    76003-308

Specifications

About this item

Essential standards for predictive toxicology screening.

The Screen-Well® Myotoxicity Library is a focused collection of 60 compounds with defined and diverse myotoxicity, including rhabdomyolysis, cardiomyopathy, mitrochondrial myopathy, vacuolar myopathy, and more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. All compounds are dissolved in DMSO at 10mM unless otherwise stated in the documentation provided with the library. Selected compounds are dissolved in DI water. All compounds are provided at 100µL volumes in a 96-Well plate.Developed in collaboration with Cytoo, a life sciences CRO focused on innovative products for the cosmetics and drug discovery markets with an emphasis in physiological cell-based assays compatible with High Content Screening, this library is an essential cost-effective tool for predictive toxicology screening and assay development.

Ordering information: Contains 60 compounds with defined and diverse myotoxicity. A variety of structurally and mechanistically different compound classes, as well as nontoxic controls, are included.