Screen-Well® Wnt Pathway Library, Enzo Life Sciences
Compounds are highly relevant; many are unique.
The Screen-Well® Wnt Pathway Library v 2.0 is a focused collection of 71 compounds with defined and diverse Wnt pathway activity, including activators and inhibitors of Wnts, Dishevelled, GSK-3β and other Wnt pathway kinases, TCF/β-catenin, DKK, LRP, Axin, Porcupine, and more. Compounds are dissolved in DMSO at 10mM or 1mM and aliquoted into deep-well plates at 100 or 500µL per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-Wnt pathway molecules in cells as well as for use in in vitro applications.
: Contains 71 compounds with defined and diverse Wnt pathway activity. A variety of structurally and mechanistically different compound classes are included.
- Concentration:DMSO solutions (10mM) except for Bafilomycin A which is provided at 1mM in DMSO
Frequently Bought Together