B.T.B. Lactose Agar Dehydrated Culture Media, HiMedia
95022-372
: HiMedia
:
- Description:B.T.B. Lactose Agar
- Cat. No.:95022-372
- Supplier no.:M861-500G
For the isolation of pathogenic staphylococci.
- Microbiological culture media
- Packaged in 500 g poly bottles