Order Entry
United States
ContactUsLinkComponent
De-Slagging Hoods
De-Slagging Hoods
Catalog # 11022-168
undefined
De-Slagging Hoods
Catalog # 11022-168
Supplier Number:  107239-07A

Specifications

  • Description:
    De-Slagging Dust Hood
  • Outside height:
    78"
  • Outside depth:
    40"
  • Outside width:
    72"
  • Cat. No.:
    11022-168
  • Weight:
    1025 lbs. (total)
  • Supplier no.:
    107239-07A

Specifications

About this item

A six-foot wide dust hood complete with discharge chute. Slagging hoods effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants. These slagging hoods have a convenient steel post with a steel pad to hammer your lead buttons to remove the slag. There is also a chute where the slag is deposited for disposal. Designed for industrial and mining applications, our slagging hoods have a 10” flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).

  • For use with fire assays
  • Made of steel and blue powder coated for a long-lasting finish (additional colours available on request)
  • Complete with LED lighting and a compressed air hose
  • Standard size is 6ft but custom sizes are also available

After deslagging the cupels, the slag can be directed to the discharge chute to be collected in a pail for disposal.