Advanced Signal Generator
Catalog # 470229-040
Supplier: OSAW INDUSTRIAL PRODUCTS SE
Specifications
- Description:Advanced Signal Generator
- Cat. No.:470229-040
Specifications
About this item
High power and versatility.
- Low distortion with four decade ranges
- Produces square, sine, triangle, and TTL waveforms
- Built-in speaker
- Equipped with provisions for AM and FM modulation
- A weak external signal can also be amplified through the unit
Ideal for general-purpose applications in a student laboratory, particularly regarding wave phenomena. Unit has an output range of 10Hz to 110KHz, in four decade ranges.