Order Entry
United States
ContactUsLinkComponent
Brilliant Green 2%-Bile Broth Dehydrated Culture Media, MilliporeSigma
Brilliant Green 2%-Bile Broth Dehydrated Culture Media, MilliporeSigma
Catalog # EM1.05454.0500
Supplier:  MilliporeSigma
CAS Number:  
undefined
Brilliant Green 2%-Bile Broth Dehydrated Culture Media, MilliporeSigma
Catalog # EM1.05454.0500
Supplier:  MilliporeSigma
Supplier Number:  1.05454.0500
CAS Number:  

Specifications

  • Description:
    Brilliant Green 2%-Bile Broth
  • Size:
    500 g
  • Cat. No.:
    EM1.05454.0500
  • Supplier no.:
    1.05454.0500

Specifications

About this item

Media is granulated.

  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media

Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.