BD BBL™ Selective Seven H11 Agar Prepared Media Stacker™ Plates, BD Diagnostics
Catalog # 90001-560
Supplier: Avantor
CAS Number:
Specifications
- Description:Selective Seven H11 Agar (Deep Fill)
- Cat. No.:90001-560
- Supplier no.:221868
Specifications
About this item
Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100 x 10 mm or 100 x 15 mm petri-style configurations.
- Plates contain a single medium
- Interlocking, stackable design to minimize sliding or sample disruption
- Ready for immediate use
Store at 2 to 8 °C (36 to 46 °F).