BD BBL™ Sabouraud Dextrose Agar with Chloramphenicol and Gentamicin Prepared Media Stacker™ Plates, BD Diagnostics
90008-424
: Avantor
:
- Description:Sabouraud Dextrose Agar, with Chloramphenicol and Gentamicin (Deep Fill)
- Cat. No.:90008-424
- Supplier no.:296359
Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100 x 10 mm or 100 x 15 mm petri-style configurations.
- Plates contain a single medium
- Interlocking, stackable design to minimize sliding or sample disruption
- Ready for immediate use
Store at 2 to 8 °C (36 to 46 °F).