Order Entry
United States
ContactUsLinkComponent
BD BBL™ Stacker™ Mueller Hinton Agar with 5% Sheep Blood Prepared Media Stacker™ Plates, BD Diagnostics
BD BBL™ Stacker™ Mueller Hinton Agar with 5% Sheep Blood Prepared Media Stacker™ Plates, BD Diagnostics
Catalog # 90006-179
Supplier:  BD
CAS Number:  
BD BBL™ Stacker™ Mueller Hinton Agar with 5% Sheep Blood Prepared Media Stacker™ Plates, BD Diagnostics
Catalog # 90006-179
Supplier:  BD
Supplier Number:  221176
CAS Number:  

Specifications

  • Description:
    Mueller Hinton Agar, with 5% Sheep Blood
  • Cat. No.:
    90006-179
  • Supplier no.:
    221176

Specifications

About this item

Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100 x 10 mm or 100 x 15 mm petri-style configurations.

  • Plates contain a single medium
  • Interlocking, stackable design to minimize sliding or sample disruption
  • Ready for immediate use

Store at 2 to 8 °C (36 to 46 °F).