Terrific Broth Complete with Animal-Free Soytone Bottled Media, Sterile, Teknova
Catalog # 76423-620
Supplier: Teknova
CAS Number:
Specifications
- Description:Terrific broth complete with animal-free soytone
- Sterility:Sterile
- Cat. No.:76423-620
- Volume:1 L
Specifications
About this item
For the growth of recombinant E.coli
- No animal-based components
- Sterile
Due to the enriched formulation of TB, strains have an extended growth phase and this results in high cell yields. TB is commonly used for plasmid purification and protein production. Ingredients: 2% animal-free soytone (20 g/L), 2.4% yeast xxtract (24.0 g/L), 0.4% glycerol (4.0 g/L), 72 mM dipotassium phosphate, 17 mM mono potassium phosphate. Ambient shipping conditions. Store at room temperature.