ContactUsLinkComponent
Terrific Broth Complete, Bottled Media, Teknova
Terrific Broth Complete, Bottled Media, Teknova
Catalog # 100219-866
Supplier:  Teknova
CAS Number:  
Terrific Broth Complete, Bottled Media, Teknova
Catalog # 100219-866
Supplier:  Teknova
Supplier Number:  T7060
CAS Number:  
Specifications
  • Description:
    Terrific Broth Complete
  • Cat. No.:
    100219-866
  • Volume:
    1 L
  • Supplier no.:
    T7060
Specifications
About this item
Used to obtain high yields of plasmid DNA from E.coli.
  • 500 mL and 1 Liter
  • Sterile
Contains 1.2% Tryptone, 2.4% yeast extract, 0.4% glycerol, 72 mM K2HPO4, and 17 mM KH2PO4. Ambient shipping conditions. Store at room temperature.