ContactUsLinkComponent
Applicator Forceps, Locking, Sklar
Applicator Forceps, Locking, Sklar
Catalog # 75996-290
Supplier:  Sklar
undefined
Applicator Forceps, Locking, Sklar
Catalog # 75996-290
Supplier:  Sklar
Supplier Number:  98-7654
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    140 mm (5½")
  • Material:
    Stainless steel
  • Curvature:
    Straight
  • Cat. No.:
    75996-290
Specifications
About this item
Locking applicator forceps.
  • Reusable
  • Nonsterile
For sizes 1, 2, and 3 clamps.