ContactUsLinkComponent
Mehdorn Applying Forceps, Sklar
Mehdorn Applying Forceps, Sklar
Catalog # 75996-218
Supplier:  Sklar
undefined
Mehdorn Applying Forceps, Sklar
Catalog # 75996-218
Supplier:  Sklar
Supplier Number:  98-4214
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Material:
    Stainless steel
  • Tip configuration:
    Blunt
  • Curvature:
    Straight
  • Cat. No.:
    75996-218
Specifications
About this item
Sklar's® Medhorn forceps are used to place micro vessel clamps in position during surgical procedures to reduce blood flow.
  • Without lock
  • Reusable and nonsterile
Unlike the Biemer, the Medhorn is not available with a locking catch.