ContactUsLinkComponent
Littauer Wireform Scissors, Disposable, Sterile, Sklar
Littauer Wireform Scissors, Disposable, Sterile, Sklar
Catalog # 75996-104
Supplier:  Sklar
Littauer Wireform Scissors, Disposable, Sterile, Sklar
Catalog # 75996-104
Supplier:  Sklar
Supplier Number:  96-8973
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    11.4 cm (4½")
  • Material:
    Stainless steel
  • Cat. No.:
    75996-104
  • Sterility:
    Sterile
Specifications
About this item
Econo™ Sterile Littauer Wireform Scissors are a ready-to-use, sterile, disposable, floor-grade instrument.
  • Small hook-shaped tip on one blade
  • Hook holds the suture so it does not slip off blade
These scissors are primarily used for suture removal. The finger ring scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They should not be used for cutting tissue.