ContactUsLinkComponent
Wireform Thumb Forceps, Disposable, Sterile, Sklar
Wireform Thumb Forceps, Disposable, Sterile, Sklar
Catalog # 75996-096
Supplier:  Sklar
undefined
Wireform Thumb Forceps, Disposable, Sterile, Sklar
Catalog # 75996-096
Supplier:  Sklar
Supplier Number:  96-8963
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    11.4 cm (4½")
  • Material:
    Stainless steel
  • Tip configuration:
    1 x 2 Teeth
  • Cat. No.:
    75996-096
  • Sterile:
    +
Specifications
About this item
Wireform thumb forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures.
  • Multipurpose
  • Sterile
  • Toothed
This product is straight with 1x2 teeth, and a length of 4-1/2 inches. This sterile, disposable instrument is sold in boxes of 50.