Specifications
- Description:Minimal M9 broth
- Sterility:Sterile
- Cat. No.:76423-596
- Volume:1 L
Specifications
About this item
For the maintenance and propagation of E.coli.
- 1 Liter
- Sterile
Contains 1× M9 salts (42 mM Na₂HPO₄, 24 mM KH₂PO₄, 9 mM NaCl, 19 mM NH₄Cl), 1 mM MgSO₄, 0.1 mM CaCl₂, 2.0% glucose, 0.5 µg/ml thiamine. Ambient shipping conditions. Store at room temperature.