BCR® Flat Mop Holder, Berkshire
Catalog # 76028-244
Supplier: Berkshire
Specifications
- Description:"V" Flat Mop Holder
- Material:Polyethylene
- Environmentally Preferable:
- Cat. No.:76028-244
Specifications
About this item
The Ergonomic 16" Flat Mop Holder is a versatile flat mop that allows cleanroom staff can attach mops faster and more efficiently than ever. Due to the "V" shape, it slips smoothly into the pouches of the mop, reducing bending and decreasing injury.
- Ergonomic holder
- Lightweight
- Detachable wiper clips
- Autoclavable
Easy insertion of the holder into the pouches of the mop is possible due to the unique "V" shape
Lightweight design makes it ideal for cleaning walls and ceilings. Extremely maneuverable and smooth-running.
An ergonomic hand-switch opens the holder and allows mop discarding while standing upright, so there is no contact with soiled mops when discarding.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.