ContactUsLinkComponent
BCR® Cleanroom String Mop Wringer with Handle, Berkshire
BCR® Cleanroom String Mop Wringer with Handle, Berkshire
Catalog # 76028-250
Supplier:  Berkshire
undefined
BCR® Cleanroom String Mop Wringer with Handle, Berkshire
Catalog # 76028-250
Supplier:  Berkshire
Supplier Number:  BCRSTRWRINGER
Specifications
  • Description:
    String Mop Wringer with Handle
  • Material:
    Durable Plastic
  • Cat. No.:
    76028-250
Specifications
About this item
The Cleanroom string mop wringer is made of durable plastic, and is suitable for the one-step wet cleaning process.
  • Durable plastic construction
  • Ergonomic handle
  • ISO Class 3–8
  • Cleanroom Class 1–100,000
Wringer is made of high-quality durable plastic, suitable for the one-step wet cleaning process. Ergonomically designed handle conveniently serves as a rest for the mop handle while the mop is not in use.