Order Entry
United States
ContactUsLinkComponent
Glutaraldehyde 10% in methanol, anhydrous, EM Grade
Glutaraldehyde 10% in methanol, anhydrous, EM Grade
Catalog # 100504-018
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Glutaraldehyde 10% in methanol, anhydrous, EM Grade
Catalog # 100504-018
Supplier Number:  16532
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10×10 ml
  • Cat. No.:
    100504-018
  • Supplier no.:
    16532

Specifications

About this item

 :
 :100.12
UN:2927
ADR:6.1,II
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025