Lead Standard Solution, for Atomic Grade Spectroscopy, PerkinElmer
Catalog # 97012-012
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:2% HNO₃
- Size:125 ml
- Cat. No.:97012-012
- Element:Lead
- Supplier no.:N9300175
Specifications
About this item
Pure Grade Single element Standards for AA and ICP-OES.
- Matrix of 2% HNO3
- Concentration of 1000 µg/mL
Certifications: ISO 17034 and ISO 17025, certified by A2LA
Delivery: Supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.