Anti-ZNF21 Rabbit Polyclonal Antibody
102030-954
: Avantor
- Antibody Type:Primary
- Antigen Symbol:ZNF21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western Blot:Yes
- Size:50 µg
- Format:Lyophilized powder. Add 50 ul of distilled water. Final antibody concentration is 1 mg/ml in PBS buffer.
- Storage Temperature:Store at 4 deg C, following reconstitution, aliquot and store at -20 deg C. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:ZNF21 antibody was raised in rabbit using the middle region of ZNF21 as the immunogen
- Cat. No.:102030-954
- Supplier no.:70R7993
Located on chromosome X, ZNF21 encodes a zinc finger protein 21.
WB: 0.2-1 ug/ml
Type: Primary
Antigen: ZNF21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: