To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:VWF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Dog
- Size:5 mg
- Format:Supplied in liquid form with 10 mM HEPES, pH 7.4, 150 mM NaCl, 50% (v/v) glycerol.
- Storage Temperature:Store at -10 to -20 deg C. Product will become viscous but will not freeze. Avoid storage in frost-free freezers. Allow product to warm to room temperature and gently mix before use.
- Shipping Temperature:*Blue Ice*
- Immunogen:VWF antibody was raised in sheep using Canine vWF purified from plasma as the immunogen.
- Cat. No.:101987-738
- Supplier no.:20RVS002
Specifications
About this item
Von Willebrand factor (vWF) is a blood glycoprotein involved in hemostasis. It is deficient or defective in von Willebrand disease and is involved in a large number of other diseases, including thrombotic thrombocytopenic purpura, Heyde's syndrome, and possibly hemolytic-uremic syndrome.
Type: Primary
Antigen: VWF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Dog