To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:E. coli (K99 Pilus)
- Clonality:Monoclonal
- Clone:M2110124
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:E. coli
- Size:500 µg
- Environmentally Preferable:
- Format:Purified IgG, supplied in PBS buffer, pH 7.4 with no preservatives added.
- Storage Temperature:Aliquot and store at -70 deg C or lower. Avoid repeated freeze/thaw cycles
- Concentration:Batch dependent - please inquire should you have specific requirements
- Shipping Temperature:*Blue Ice*
- Immunogen:E. coli antibody was raised in mouse using purified E. coli K99 pili as the immunogen.
- Cat. No.:101977-762
- Supplier no.:10E20A
Specifications
About this item
Escherichia coli (commonly abbreviated E. coli, named after Theodor Escherich) is a Gram-negative rod-shaped bacterium that is commonly found in the lower intestine of warm-blooded organisms (endotherms). Most E. coli strains are harmless, but some, such as serotype O157:H7, can cause serious food poisoning in humans, and are occasionally responsible for product recalls. The harmless strains are part of the normal flora of the gut, and can benefit their hosts by producing vitamin K2, and by preventing the establishment of pathogenic bacteria within the intestine.
ELISA: 1:102,400
Type: Primary
Antigen: E. coli (K99 Pilus)
Clonality: Monoclonal
Clone: M2110124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: E. coli