Anti-ACOT11 Mouse Monoclonal Antibody [clone: J4B2]
Catalog # 101978-704
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Symbol:ACOT11
- Clonality:Monoclonal
- Clone:J4B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Format:Supplied as a liquid in PBS, pH 7.4, with 0.1% NaN3.
- Storage Temperature:Store at 4 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Concentration:1 mg/ml
- Immunogen:ACOT11 antibody was raised in mouse using recombinant human ACOT11 (19-250aa) purified from E. coli as the immunogen.
- Cat. No.:101978-704
- Supplier no.:10R1102
Specifications
About this item
ACOT11 (Acyl-CoA thioesterase11), also known as BFIT, is a protein with acyl-CoA thioesterase activity towards medium (C12) and long-chain (C18) fatty acyl-CoA substrates. Expression of this protein in mouse has been associated with obesity and alternative splicing results in two transcript variants encoding different isoforms.
IHC: 1:50-1:100, WB: 1:500-1:2,000
Type: Primary
Antigen: ACOT11
Clonality: Monoclonal
Clone: J4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human