Borosilicate Glass VOA Vials, Sun Sri
Supplier: Thermo Scientific
Certificates
About this item
VOA vials are made from clear borosillicate glass, and are used for the analysis of volatile organic compounds.
Vials have an operational temperature range from –40 to 125 °C (–40 to 257 °F).
Ordering information: Vials, caps, and septa are available as separate components or as kits.
Frequently Bought Together