ContactUsLinkComponent
WesternSure™ Pen, LI-COR®
WesternSure™ Pen, LI-COR®
Catalog # 102971-112
Supplier:  Li-Cor
undefined
WesternSure™ Pen, LI-COR®
Catalog # 102971-112
Supplier:  Li-Cor
Supplier Number:  926-91000
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    WesternSure™ Pen
  • Cat. No.:
    102971-112
  • Supplier no.:
    926-91000
Specifications
About this item
The WesternSure Pen is used to annotate visible protein ladders prior to chemiluminescent Western blot detection.
  • Optimized for detection using the C-DiGit® Blot Scanner
  • Optimized for detection using the Odyssey® Fc Imaging System
A unique marker is used which delivers an ink which emits light when incubated with commonly-used chemilumi­nescent substrates, including WesternSure PREMIUM
The WesternSure Pen was used to mark the blue protein standards for chemiluminescent Western blot detection. The blot was exposed to WesternSure PREMIUM chemiluminescent substrate and imaged on Odyssey Fc Imaging System.
The ink is faintly visible for easy identification of marked membranes.