Order Entry
United States
ContactUsLinkComponent
L(+)-Aspartic acid, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
L(+)-Aspartic acid, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Supplier:  MilliporeSigma
Certificates
L(+)-Aspartic acid, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Supplier:  MilliporeSigma
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Assay (alkalimetric, calculated on dried substance):
    98.5 - 101.5 %
  • Identity (IR-spectrum):
    passes test
  • Identity (enantiomeric purity):
    passes test
  • Appearance:
    white to almost white, crystalline powder or colorless crystals
  • Appearance of solution (50 g/l; hydrochloric acid 1 mol/l):
    clear and not more intense in color than reference solution BY₆
  • Spec. rotation (α 20/D, 80 g/l, hydrochloric acid 6 mol/l, calc. on dried substance):
    +24.0 to +26.0
  • Chloride (Cl):
    ≤ 200 ppm
  • Sulfate (SO₄):
    ≤ 300 ppm
  • Fe (Iron):
    ≤ 10 ppm
  • Enantiomeric purity (HPLC) (D-aspartic acid):
    ≤ 0.3 %
  • Dicarboxylic acids (HPLC) (malic acid):
    ≤ 0.2 %
  • Dicarboxylic acids (HPLC) (fumaric acid):
    ≤ 0.10 %
  • Dicarboxylic acids (HPLC) (maleic acid):
    ≤ 0.10 %
  • Dicarboxylic acids (HPLC) (any unspecified impurity):
    ≤ 0.10 %
  • Dicarboxylic acids (HPLC) (total unspecified impurities):
    ≤ 0.3 %
  • Related compounds (HPLC) (maleic acid):
    ≤ 0.05 %
  • Related compounds (HPLC) (malic acid):
    ≤ 0.20 %
  • Related compounds (HPLC) (fumaric acid):
    ≤ 0.10 %
  • Related compounds (HPLC) (any unspecified impurity):
    ≤ 0.05 %
  • Related compounds (HPLC) (total unspecified impurities):
    ≤ 0.10 %
  • Ninhydrin-positive substances (LC)(impurity C (glutamic acid)):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC)(impurity D (alanine)):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC)(impurity G (asparagine)):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC) (any ninhydrin-positive impurity):
    ≤ 0.10 %
  • Ninhydrin-positive substances (LC) (total impurities):
    ≤ 1.0 %
  • Ninhydrin-positive substances (LC) (ammonium (570 nm)):
    ≤ 0.02 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Loss on drying (105 °C; 3 h):
    ≤ 0.5 %

Specification Test Results

Related Information

Frequently Bought Together