L(+)-Tartaric acid Ph. Eur., JP, NF, BP endotoxin tested
Supplier: Avantor Performance Materials
Certificates
About this item
Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
CAS Number:87-69-4
:
Formula:C₄H₆O₆
MDL Number:MFCD00064207
Boiling Pt:399
Molecular Weight:150.09 g/mol
Flash Pt:150
Merck Index:13,09158
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (NF):99.7 - 100.5%
- Bacterial Endotoxins (NF, EP, BP, JP):≤ 10 EU/g
- Heavy Metals 231:≤ 10 ppm
- Identification A (NF, EP, BP):Pass
- Identification B (NF, EP, BP):Pass
- Limit of Oxalate Test (NF):Pass
- Loss on Drying 731:≤ 0.5%
- Residue on Ignition 281:≤ 0.1%
- Specific Rotation 781s:+12.0° to +13.0°
- Sulfate Test:Pass
- Appearance of Solution (EP, BP):Pass
- Assay (dried substance, EP, BP):99.5 - 101.0%
- Calcium (2.4.3):≤ 200 ppm
- Chlorides (2.4.4):≤ 100 ppm
- Loss on Drying (2.2.32):≤ 0.2%
- Oxalic Acid:≤ 360 ppm
- Specific Optical Rotation (2.2.7):+12.0° to +12.8°
- Sulfated Ash (2.4.14):≤ 0.1%
- Sulfates (2.4.13):≤ 150 ppm
- Arsenic 1.11:≤ 1 ppm
- Assay (dried, JP):≥ 99.7%
- Calcium Test (JP):Pass
- Heavy Metals 1.07:≤ 10 ppm
- Identification (1) (JP):Pass
- Identification (2) (JP):Pass
- Loss on Drying ,2.41:≤ 0.5%
- Oxalate Test (JP):Pass
- Residue on Ignition 2.44:≤ 0.05
- Sulfate 1.14:≤ 0.048 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberL-TARTARIC ACID VWRB501 Product SpecificationsREV2.pdf spec_VWRB501-500G spec_VWRB501-500G spec_VWRB501-50KG
Product specification
VWR Bioprocessing Chemical and Excipients
Product Literature
Frequently Bought Together