FAME #1 Mix (5 components), Restek
Catalog # RK35010
Supplier: Restek
CAS Number:
Specifications
- Concentration:Neat
- Description:Fatty acid methyl ester, FAME Std #1
- Size:50 mg
- Cat. No.:RK35010
- Supplier no.:35010
Specifications
About this item
Contains: Chain, Compound (CAS#), % by Weight; C16:00 Methyl palmitate (112-39-0), 20%; C18:00 Methyl stearate (112-61-8), 20%; C18:01 Methyl oleate (112-62-9), 20%; C18:02 Methyl linoleate (112-63-0), 20%; C18:03 Methyl linolenate (301-00-8), 20%