Order Entry
United States
ContactUsLinkComponent
PCB Kit #1, Restek
PCB Kit #1, Restek
Catalog # RK32089
Supplier:  Restek
CAS Number:  
undefined
PCB Kit #1, Restek
Catalog # RK32089
Supplier:  Restek
Supplier Number:  32089
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in hexane
  • Description:
    PCB Kit #1
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK32089
  • Certification:
    CRM
  • Supplier no.:
    32089

Specifications

About this item

This kit contains Aroclor 1016, Aroclor 1221, Aroclor 1232, Aroclor 1242, Aroclor 1248, Aroclor 1254, Aroclor 1260.

  • Aroclor standards may exhibit lot-to-lot variation.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.