Order Entry
United States
ContactUsLinkComponent
Herbicide Mix #3 (2 components), Derivatized Form, Restek
Herbicide Mix #3 (2 components), Derivatized Form, Restek
Catalog # RK32059
Supplier:  Restek
CAS Number:  
undefined
Herbicide Mix #3 (2 components), Derivatized Form, Restek
Catalog # RK32059
Supplier:  Restek
Supplier Number:  32059
CAS Number:  

Specifications

  • Concentration:
    20000 µg/ml each in hexane
  • Description:
    Herbicide Mix #3
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK32059
  • Certification:
    CRM
  • Supplier no.:
    32059

Specifications

About this item

This mix contains two components.

Components are MCPA methyl ester, MCPP (mecoprop) methyl ester.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.