WA EPH Aromatic Hydrocarbon Standard (18 Components), Restek
Catalog # RK31469
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in methylene chloride
- Description:WA EPH Aromatic Hydrocarbon Standard
- Size:1 ml/Ampule
- Cat. No.:RK31469
- Certification:CRM
- Supplier no.:31469
Specifications
About this item
Mix contains: Acenaphthene (83-32-9); Acenaphthylene (208-96-8); Anthracene (120-12-7); Benz(a)anthracene (56-55-3); Benzo(a)pyrene (50-32-8); Benzo(b)fluoranthene (205-99-2); Benzo(k)fluoranthene (207-08-9); Benzo(ghi)perylene (191-24-2); Chrysene (218-01-9); Dibenz(a,h)anthracene (53-70-3); Fluoranthene (206-44-0); Fluorene (86-73-7); Indeno(1,2,3-cd)pyrene (193-39-5); 2-Methylnaphthalene (91-57-6); Naphthalene (91-20-3); Phenanthrene (85-01-8); Pyrene (129-00-0); 1,2,3-Trimethylbenzene (526-73-8).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.