612 Chlorinated Hydrocarbons Calibration Mix (9 components), Restek
Catalog # RK31035
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in isooctane
- Description:612 Chlorinated Hydrocarbons Calibration Mix
- Size:1 ml/Ampule
- Cat. No.:RK31035
- Certification:CRM
- Supplier no.:31035
Specifications
About this item
612 Chlorinated hydrocarbons calibration mix contains 2-chloronaphthalene, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, hexachlorobenzene, hexachlorobutadiene, hexachlorocyclopentadiene, hexachloroethane, 1,2,4-trichlorobenzene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.