ContactUsLinkComponent
611 Haloethers Calibration Mix (5 components), Restek
611 Haloethers Calibration Mix (5 components), Restek
  RK31034
 :  Restek
 :  
undefined
611 Haloethers Calibration Mix (5 components), Restek
  RK31034
 :  Restek
 :  31034
 :  
  • Concentration:
    2000 µg/ml each in acetone
  • Description:
    611 Haloethers Calibration Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK31034
  • Certification:
    CRM
  • Supplier no.:
    31034
Haloethers calibration mix contains bis(2-chloroethoxy)methane, bis(2-chloroethyl)ether, 4-bromophenyl phenyl ether, 4-chlorophenyl phenyl ether, 2,2'-oxybis(1-chloropropane).
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.